$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Stigmasteryl Ferulate
TRC Stigmasteryl Ferulate
77993-471
:
LGC Standards
New Product
:
TRC Stigmasteryl Ferulate
77993-471
:
LGC Standards
:
TRC-S686768-2.5MG
New Product
:
CAS No.:
20972-08-1
Description:
Stigmasteryl Ferulate
Size:
2.5 mg