Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC N-Desmethyl Nicergoline
TRC N-Desmethyl Nicergoline
TRC N-Desmethyl Nicergoline
77982-268
:
LGC Standards
New Product
:
TRC N-Desmethyl Nicergoline
77982-268
:
LGC Standards
:
TRC-D422360-10MG
New Product
:
CAS No.:
192504-81-7
Description:
N-Desmethyl Nicergoline
Size:
10 mg