$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Haloperidol
TRC Haloperidol
Catalog # 77985-799
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Haloperidol
Catalog # 77985-799
Supplier:
LGC Standards
Supplier Number:
TRC-H103700-100MG
New Product
CAS Number:
Specifications
CAS No.:
52-86-8
Description:
Haloperidol
Size:
100 mg
Specifications
About this item
Product Details
Product Details