$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Glyoxal (40% w/w in H2O)
TRC Glyoxal (40% w/w in H2O)
77985-704
:
LGC Standards
New Product
:
TRC Glyoxal (40% w/w in H2O)
77985-704
:
LGC Standards
:
TRC-G754500-25ML
New Product
:
Matrix:
Water
CAS No.:
107-22-2
Description:
Glyoxal (40% w/w in H2O)
Size:
25 ml