Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Nitrate Visual Test Kit Zinc Reduction
CHEMets® Nitrate Visual Test Kit Zinc Reduction
Catalog # 10736-448
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Nitrate Visual Test Kit Zinc Reduction
Catalog # 10736-448
Supplier:  CHEMetrics
Supplier Number:  K-6905
New Product
CAS Number:  

Specifications

  • Chemical method:
    Zinc reduction
  • Cat. No.:
    10736-448
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Boiler water;Cooling water;Surface and ground water;Potable water
  • Range:
    0 - 3.4 ppm as N
  • No. of tests:
    30

Specifications

About this item

  • Range: 0 to 3.4 ppm as N MDL: 0.3 ppm Method: Zinc reduction
Delivery: Nitrate test kit comes in a plastic case and contains everything needed to perform 30 tests:refill, comparator, acidifier solution, reaction tube and cap, 25 ml sample cup, and instructions.