Paragon Scientific Sulfur @ 0.25% (2500 ppm) in odorless kerosene
77975-838
New Product
:
- Matrix:Odourless Kerosene
- CAS No.:7704-34-9
- Description:Sulfur @ 0.25% (2500 ppm) in odorless kerosene
- Size:100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065