TRC 4-(4-Morpholinyl)-1,2,5-thiadiazol-3(2H)-one 1-Oxide
New Product
- CAS No.:75202-36-7
Frequently Bought Together
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
: Antyila Scientific
CrossLab Instrument Supplies for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Flame Ionization Detector (FID) for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for Gas Clean Purification System, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Closures for 2 ml Screw Top Vials, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
CrossLab Drain Tubing for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Baskets, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Replacement Frits for Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sample Loops for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
InfinityLab Well Plates, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
O-Rings and Fittings for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
ZORBAX, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)