Lipomed d,l-MDEA-D5.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-199
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:1286588-92-8
- Description:d,l-MDEA-D5.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml