TRC Methyl 3,4,5-Trimethoxybenzoate
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:1916-07-0
Specifications
Frequently Bought Together
CrossLab Headspace Sampler Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Baskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fraction Collector Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
Accessories for Gas Clean Purification System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Purged Packed Inlet for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® PFA and PFA HP Fluoropolymer Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
CrossLab Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for VacElut Cartridge Manifolds, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)