TRC Caffeine
New Product
- CAS No.:58-08-2
Frequently Bought Together
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
: Antyila Scientific
Captiva Syringe Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Sample Tubes and Vials, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Replacement Frits for Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Bond Elut™ Adapters and Caps, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for Stay Safe Caps for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Gas Tubing for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
Guard Cartridge Holder, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Nebulizer Supplies for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Pump Valves and Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fraction Collector Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)