Lipomed d,l-Amphetamine.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-094
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:2706-50-5
- Description:d,l-Amphetamine.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml