Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 1-Naphthylacetyl Spermine Trihydrochloride
TRC 1-Naphthylacetyl Spermine Trihydrochloride
TRC 1-Naphthylacetyl Spermine Trihydrochloride
77990-006
:
LGC Standards
New Product
:
TRC 1-Naphthylacetyl Spermine Trihydrochloride
77990-006
:
LGC Standards
:
TRC-N377800-10MG
New Product
:
CAS No.:
1049731-36-3
Description:
1-Naphthylacetyl Spermine Trihydrochloride
Size:
10 mg