Paragon Scientific ASTM D2622, Sulfur set B in light mineral oil: S @ 0.1, 0.25, 0.5, 1%
77975-599
New Product
:
- Matrix:Light Mineral Oil
- CAS No.:7704-34-9
- Description:ASTM D2622, Sulfur set B in light mineral oil: S @ 0.1, 0.25, 0.5, 1%
- Size:4×100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065