Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC rac 1-Oleoyl Glycerol
TRC rac 1-Oleoyl Glycerol
77990-829
:
LGC Standards
New Product
:
TRC rac 1-Oleoyl Glycerol
77990-829
:
LGC Standards
:
TRC-O527980-250MG
New Product
:
CAS No.:
111-03-5
Description:
rac 1-Oleoyl Glycerol
Size:
250 mg