TRC Pyridoxal 5'-Phosphate
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:54-47-7
Specifications
Frequently Bought Together
CrossLab Dissolution Baskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Nebulizers for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® ETFE Fluoropolymer Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
VacElut Cartridge Manifolds, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for In-line Gas Purifiers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
O-Rings and Fittings for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)