Paragon Scientific ASTM D4294, Sulfur set C in odorless kerosene: S @ 1, 2, 3, 4, 5%
Catalog # 77975-608
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Odourless Kerosene
- CAS No.:7704-34-9
- Description:ASTM D4294, Sulfur set C in odorless kerosene: S @ 1, 2, 3, 4, 5%
- Size:5×100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065