TRC Imipramine Hydrochloride
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:113-52-0
Specifications
Frequently Bought Together
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Macrocells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for HPLC Columns, Hypersil, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Purged Packed Inlet for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Gas Tubing for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Stay Safe Caps & Accessories for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab GC Fittings, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Sealing Mats for InfinityLab Well Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Fused Silica Tubing Cutters for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)