Paragon Scientific Sulphur in diesel, low (7.2 mg/kg)
77975-560
New Product
:
- Matrix:Diesel
- CAS No.:7704-34-9
- Description:Sulphur in diesel, low (7.2 mg/kg)
- Size:250 ml
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065