$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Metoclopramide N-Oxide
TRC Metoclopramide N-Oxide
:
LGC Standards
New Product
TRC Metoclopramide N-Oxide
:
LGC Standards
New Product
CAS No.:
171367-22-9