Paragon Scientific IP 336, Sulfur Set D in light mineral oil: S @ 2, 3, 4, 5%
Catalog # 77975-816
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Light Mineral Oil
- CAS No.:7704-34-9
- Description:IP 336, Sulfur Set D in light mineral oil: S @ 2, 3, 4, 5%
- Size:4×100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065