$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC L-Methionine-d3
TRC L-Methionine-d3
TRC L-Methionine-d3
Supplier:
LGC Standards
New Product
Certificates
Expand image
TRC L-Methionine-d3
Supplier:
LGC Standards
New Product
About this item
Specifications
CAS No.:
13010-53-2
Specifications