$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Chondroitin Sulfate, Bovine Trachea
TRC Chondroitin Sulfate, Bovine Trachea
Catalog # 77980-282
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Chondroitin Sulfate, Bovine Trachea
Catalog # 77980-282
Supplier:
LGC Standards
Supplier Number:
TRC-C433045-250MG
New Product
CAS Number:
Specifications
CAS No.:
9007-28-7
Description:
Chondroitin Sulfate, Bovine Trachea
Size:
250 mg
Specifications
About this item
Product Details
Product Details