Order Entry
Puerto Rico
ContactUsLinkComponent
Chromate Comparator Round 0 to 1 ppm
Chromate Comparator Round 0 to 1 ppm
Catalog # 77674-606
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Chromate Comparator Round 0 to 1 ppm
Catalog # 77674-606
Supplier:  CHEMetrics
Supplier Number:  C-2801
New Product
CAS Number:  

Specifications

  • Chemical method:
    Diphenylcarbazide

Specifications

About this item

C-2801 Low Range Chromate Comparator is a component of K-2810 Chromate (hexavalent) CHEMets Visual Test Kit and is designed to be compared against the color formed in R-2810 Chromate CHEMets Refill. The comparator's range is applicable for many industrial wastewater applications. Color standards are pink in color and cover a range of 0 to 1.0 ppm CrO₄.