Lipomed Buprenorphine-D4.HCl, 0.1 mg/ml in Methanol (as free base)
Catalog # 77976-101
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- Description:Buprenorphine-D4.HCl, 0.1 mg/ml in Methanol (as free base)
- Size:1 ml