Lipomed Amitriptyline-D3.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-087
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:342611-00-1
- Description:Amitriptyline-D3.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml