Paragon Scientific IP 336, Sulfur Set F in synthetic diesel: S @ 0.5, 1, 1.5, 2, 3, 4, 5%
Catalog # 77975-800
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:7704-34-9
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065