$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Montelukast Bis-sulfide (mixture of diastereomers)
TRC Montelukast Bis-sulfide (mixture of diastereomers)
77989-743
:
LGC Standards
New Product
:
TRC Montelukast Bis-sulfide (mixture of diastereomers)
77989-743
:
LGC Standards
:
TRC-M567985-10MG
New Product
:
CAS No.:
1242260-05-4
Description:
Montelukast Bis-sulfide (mixture of diastereomers)
Size:
10 mg