Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Sulindac Sulfone Acyl-Beta-D-Glucuronide
TRC Sulindac Sulfone Acyl-Beta-D-Glucuronide
TRC Sulindac Sulfone Acyl-Beta-D-Glucuronide
77993-665
:
LGC Standards
New Product
:
TRC Sulindac Sulfone Acyl-Beta-D-Glucuronide
77993-665
:
LGC Standards
:
TRC-S699270-1MG
New Product
:
CAS No.:
60018-37-3
Description:
Sulindac Sulfone Acyl-Beta-D-Glucuronide
Size:
1 mg