Order Entry
Puerto Rico
ContactUsLinkComponent
 
CHEMets® Silica Test Kit (0 to 1 and 1 to 10 ppm)
Catalog # 11027-212
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Silica Test Kit (0 to 1 and 1 to 10 ppm)
Catalog # 11027-212
Supplier:  CHEMetrics
Supplier Number:  K-9010
New Product
CAS Number:  

Specifications

  • Chemical method:
    Heteropoly blue
  • Application:
    Boiler water;Cooling water;Natural water;Waste water;High purity water

Specifications

About this item

  • Range: 0 to 1 and 1 to 10 ppm MDL: 0.05 ppm Method: Heteropoly blue
Delivery: Silica kit (K-9010) comes in a plastic case and contains everything needed to perform 30 tests:refill, low and high range comparators, neutralizer solution, activator solution, 25 ml sample cup, sample cup top, and instructions.