$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Tris(1,3-dichloro-2-propyl) Phosphate
TRC Tris(1,3-dichloro-2-propyl) Phosphate
Catalog # 77994-893
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Tris(1,3-dichloro-2-propyl) Phosphate
Catalog # 77994-893
Supplier:
LGC Standards
Supplier Number:
TRC-T876305-1G
New Product
CAS Number:
Specifications
CAS No.:
13674-87-8
Description:
Tris(1,3-dichloro-2-propyl) Phosphate
Size:
1 g
Specifications
About this item
Product Details
Product Details