TRC L-Arginine
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:74-79-3
Specifications
Frequently Bought Together
pH Gradient Buffers
Supplier: Thermo Scientific
Intuvo Flow Chips, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Peristaltic Pump Tubing for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing and Fittings for 8453/8454 UV Systems
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Ion Lenses for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Split Liners for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Liner O-Rings for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab GC Fittings, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
LISA Safety Waste Cap
Supplier: SCAT AMERICAS, INC.
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
SWITCHING VALVE 4 PORT RIGHT ANGLE FLOW
Supplier: ANALYTICAL SALES & SERVICE MS
Nebulizer Supplies for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Cleaning and Maintenance Supplies for GC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)