$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Vinorelbine Bitartrate
TRC Vinorelbine Bitartrate
77995-261
:
LGC Standards
New Product
:
TRC Vinorelbine Bitartrate
77995-261
:
LGC Standards
:
TRC-V315000-25MG
New Product
:
CAS No.:
125317-39-7
Description:
Vinorelbine Bitartrate
Size:
25 mg