Order Entry
Puerto Rico
ContactUsLinkComponent
 
Lipomed d,l-MDMA-D5.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-200
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Lipomed d,l-MDMA-D5.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-200
Supplier:  LGC Standards
Supplier Number:  LPM-MDM-324-HC-1LM
New Product
CAS Number:  

Specifications

  • Matrix:
    Methanol
  • CAS No.:
    2747917-97-9
  • Description:
    d,l-MDMA-D5.HCl, 1 mg/ml in Methanol (as free base)
  • Size:
    1 ml

Specifications

About this item