Order Entry
Puerto Rico
ContactUsLinkComponent
 
Paragon Scientific Small Sample Viscosity (1706 cSt @ 40 °C), Certified @ 40 and 100 °C
  77975-777
New Product
 :  
undefined
Paragon Scientific Small Sample Viscosity (1706 cSt @ 40 °C), Certified @ 40 and 100 °C
  77975-777
 :  ALK-HVS11
New Product
 :  

 

  • Description:
    Small sample viscosity (1706 cSt @ 40 °C), certified @ 40 and 100 °C
  • Size:
    60 ml
  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT
  • Certification:
    ISO 17034

 

 

  • Standard type: Single component