Order Entry
Puerto Rico
ContactUsLinkComponent
TRC (S)-5-Tert-butoxy-4-(3-((s)-1,5-di-tert-butoxy-1,5-dioxopentan-2-yl)ureido)-5-oxopentanoic Acid
TRC (S)-5-Tert-butoxy-4-(3-((s)-1,5-di-tert-butoxy-1,5-dioxopentan-2-yl)ureido)-5-oxopentanoic Acid
Catalog # 77993-989
Supplier:  LGC Standards
New Product
CAS Number:  
TRC (S)-5-Tert-butoxy-4-(3-((s)-1,5-di-tert-butoxy-1,5-dioxopentan-2-yl)ureido)-5-oxopentanoic Acid
Catalog # 77993-989
Supplier:  LGC Standards
Supplier Number:  TRC-T205165-100MG
New Product
CAS Number:  

Specifications

  • CAS No.:
    1026987-94-9
  • Description:
    (S)-5-Tert-butoxy-4-(3-((s)-1,5-di-tert-butoxy-1,5-dioxopentan-2-yl)ureido)-5-oxopentanoic Acid
  • Size:
    100 mg

Specifications

About this item