Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-(9H-Carbazol-4-yloxy)-1,2-propanediol
TRC 3-(9H-Carbazol-4-yloxy)-1,2-propanediol
TRC 3-(9H-Carbazol-4-yloxy)-1,2-propanediol
77979-436
:
LGC Standards
New Product
:
TRC 3-(9H-Carbazol-4-yloxy)-1,2-propanediol
77979-436
:
LGC Standards
:
TRC-C176060-100MG
New Product
:
CAS No.:
123119-89-1
Description:
3-(9H-Carbazol-4-yloxy)-1,2-propanediol
Size:
100 mg