Paragon Scientific ASTM D7039, Sulfur set B in synthetic diesel: S @ blank, 0.0025, 0.01, 0.05, 0.1, 0.3%
Catalog # 77975-613
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Synthetic Diesel
- CAS No.:7704-34-9
- Description:ASTM D7039, Sulfur set B in synthetic diesel: S @ blank, 0.0025, 0.01, 0.05, 0.1, 0.3%
- Size:6×100ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065