$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Urolithin C
TRC Urolithin C
77995-085
:
LGC Standards
New Product
:
TRC Urolithin C
77995-085
:
LGC Standards
:
TRC-U847015-25MG
New Product
:
CAS No.:
165393-06-6
Description:
Urolithin C
Size:
25 mg