Order Entry
Puerto Rico
ContactUsLinkComponent
Ozone Instrumental Test 0 to 0.75 ppm
Ozone Instrumental Test 0 to 0.75 ppm
Catalog # 77674-844
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Ozone Instrumental Test 0 to 0.75 ppm
Catalog # 77674-844
Supplier:  CHEMetrics
Supplier Number:  K-7433
New Product
CAS Number:  

Specifications

  • Chemical method:
    Indigo
  • Application:
    Potable water;Food and beverages;Bottled water;Pharmaceutical solutions

Specifications

About this item

  • Range: 0 to 0.75 ppm Method: Indigo
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, 25 ml sample cup, ampoule blank, and instructions. This kit requires the use of an I-2022 SAM photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell and with a 'Z dimension' of 15 mm or less.