$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Sorafenib-d4
TRC Sorafenib-d4
Catalog # 77993-331
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Sorafenib-d4
Catalog # 77993-331
Supplier:
LGC Standards
Supplier Number:
TRC-S676853-1MG
New Product
CAS Number:
Specifications
Description:
Sorafenib-d4
Size:
1 mg
Specifications
About this item
Product Details
Product Details