ContactUsLinkComponent
Ammonia Stabilizer Accessory Solution Pack for Saltwater Analysis
Ammonia Stabilizer Accessory Solution Pack for Saltwater Analysis
Catalog # 77674-534
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Ammonia Stabilizer Accessory Solution Pack for Saltwater Analysis
Catalog # 77674-534
Supplier:  CHEMetrics
Supplier Number:  A-1501
New Product
CAS Number:  
Specifications
  • Chemical method:
    Direct Nesslerization
Specifications
About this item
The A-1501 Ammonia Stabilizer Accessory Solution Pack for Saltwater Analysis is used in conjunction with the Ammonia test kits K-1503, K-1510, and K-1523 for seawater or saltwater analysis. This pack supplies enough solution to analyze approximately 100 seawater samples. Qty. per Pack: Six 20 ml bottles.