ContactUsLinkComponent
DEHA Titration Test 25 to 250 ppm
  77674-776
New Product
 :  
undefined
DEHA Titration Test 25 to 250 ppm
  77674-776
 :  K-3925
New Product
 :  
  • Chemical method:
    Ceric sulfate titrant with Ferroin indicator
  • Application:
    Boiler feed water and condensate
Test method: DEHA Titrets® test kit (25 to 250 ppm) method employs a ceric sulfate titrant and a ferroin endpoint indicator. The sample is treated with an excess of ferric iron. DEHA (N,N-Diethylhydroxylamine) reacts quantitatively with ferric iron by reducing it to the ferrous state. The resulting ferrous iron is titrated with the ceric sulfate titrant.
  • Range: 25 to 250 ppm MDL: 25 ppm Method: Ceric sulfate titrant with ferroin indicator Increments: 25, 27.5, 30, 32.5, 35, 37.5, 40, 45, 50, 62.5, 75, 87.5, 100, 125, 175, 250 ppm
Features of titrets technology: Dependable, sealed single-use titrants Fast, easy procedure, no need to count drops No burettes or other equipment necessary Perform a titration ANYWHERE
 : DEHA Titrets® test kit (25 to 250 ppm) comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules with valve assemblies, activator solution (A to 3900), titrettor (A to 0053), 25 ml sample cup (A to 0013), and instructions.