ContactUsLinkComponent
Chromate Acidifier Solution Pack
Chromate Acidifier Solution Pack
Catalog # 77674-546
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Chromate Acidifier Solution Pack
Catalog # 77674-546
Supplier:  CHEMetrics
Supplier Number:  A-2800
New Product
CAS Number:  
Specifications
  • Chemical method:
    Diphenylcarbazide
Specifications
About this item
The Chromate Acidifier Solution A-2800 accessory pack supplies enough solution to perform at least 200 tests. Qty. per Pack: six 10 ml bottles.