Lipomed d-Propoxyphene.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-233
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:1639-60-7
- Description:d-Propoxyphene.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml