$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC N-Nitroso Desloratadine
TRC N-Nitroso Desloratadine
:
LGC Standards
New Product
TRC N-Nitroso Desloratadine
:
LGC Standards
New Product
CAS No.:
1246819-22-6