$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Perphenazine Sulfoxide
TRC Perphenazine Sulfoxide
77991-501
:
LGC Standards
New Product
:
TRC Perphenazine Sulfoxide
77991-501
:
LGC Standards
:
TRC-P291105-25MG
New Product
:
CAS No.:
10078-25-8
Description:
Perphenazine Sulfoxide
Size:
25 mg