$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC L-Carnosine-d4 (N-Beta-alanyl-d4)
TRC L-Carnosine-d4 (N-Beta-alanyl-d4)
TRC L-Carnosine-d4 (N-Beta-alanyl-d4)
77979-549
:
LGC Standards
New Product
:
TRC L-Carnosine-d4 (N-Beta-alanyl-d4)
77979-549
:
LGC Standards
:
TRC-C184192-5MG
New Product
:
CAS No.:
2714339-90-7
Description:
L-Carnosine-d4 (N-Beta-alanyl-d4)
Size:
5 mg