Paragon Scientific IP 497, Sulfur set A in odorless kerosene: S @ blank, 0.005, 0.01, 0.025, 0.03, 0.05%
Catalog # 77975-825
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Odourless Kerosene
- CAS No.:7704-34-9
- Description:IP 497, Sulfur set A in odorless kerosene: S @ blank, 0.005, 0.01, 0.025, 0.03, 0.05%
- Size:6×100ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065