$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Dibromochloronitromethane
TRC Dibromochloronitromethane
Catalog # 77982-284
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Dibromochloronitromethane
Catalog # 77982-284
Supplier:
LGC Standards
Supplier Number:
TRC-D425210-10MG
New Product
CAS Number:
Specifications
CAS No.:
1184-89-0
Description:
Dibromochloronitromethane
Size:
10 mg
Specifications
About this item
Product Details
Product Details