$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 3-(2-Amino-2-oxoethyl)-5-methylhexanoic Acid
TRC 3-(2-Amino-2-oxoethyl)-5-methylhexanoic Acid
Catalog # 77977-322
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 3-(2-Amino-2-oxoethyl)-5-methylhexanoic Acid
Catalog # 77977-322
Supplier:
LGC Standards
Supplier Number:
TRC-A619045-5G
New Product
CAS Number:
Specifications
CAS No.:
181289-15-6
Description:
3-(2-Amino-2-oxoethyl)-5-methylhexanoic Acid
Size:
5 g
Specifications
About this item
Product Details
Product Details